Assay Details
| Gene Symbol: LOC4343379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789557.2 | XP_015645043.1 | 2 - 3 | 417 | 62 | |
| GenBank mRNA | AK061373.1 | - | 2 - 3 | 398 | 62 | |
| AK288667.1 | - | 2550 | 62 | |||
More Information
Set Membership: |
|

LOC4343379
Genomic Map