Assay Details
| Gene Symbol: LOC4343413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791090.2 | XP_015646576.1 | 2 - 3 | 2310 | 117 | |
| GenBank mRNA | AK067405.1 | - | 3 - 4 | 2157 | 117 | |
More Information
Set Membership: |
|

LOC4343413
Genomic Map