Assay Details
| Gene Symbol: LOC4343413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791090.2 | XP_015646576.1 | 4 - 5 | 2624 | 104 | |
| GenBank mRNA | AK067405.1 | - | 5 - 6 | 2489 | 122 | |
More Information
Set Membership: |
|

LOC4343413
Genomic Map