Assay Details
| Gene Symbol: LOC4343414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790616.1 | XP_015646102.1 | 2 - 3 | 1211 | 79 | |
| GenBank mRNA | AK071154.1 | - | 2 - 3 | 143 | 79 | |
More Information
Set Membership: |
|

LOC4343414
Genomic Map