Assay Details
| Gene Symbol: LOC4343414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790616.1 | XP_015646102.1 | 3 - 4 | 1304 | 86 | |
| GenBank mRNA | AK071154.1 | - | 3 - 4 | 236 | 86 | |
More Information
Set Membership: |
|

LOC4343414
Genomic Map