Assay Details
| Gene Symbol: LOC4344163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792552.2 | XP_015648038.1 | 10 - 11 | 1370 | 73 | |
| XM_015792553.1 | XP_015648039.1 | 10 - 11 | 1083 | 73 | ||
| GenBank mRNA | AK103870.1 | - | 10 - 11 | 1084 | 73 | |
More Information
Set Membership: |
|

LOC4344163
Genomic Map