Assay Details
| Gene Symbol: LOC4344163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792552.2 | XP_015648038.1 | 17 - 18 | 2435 | 90 | |
| XM_015792553.1 | XP_015648039.1 | 17 - 18 | 2148 | 90 | ||
| GenBank mRNA | AK058967.1 | - | 5 - 6 | 739 | 90 | |
| AK064580.1 | - | 7 - 8 | 1061 | 90 | ||
| AK103870.1 | - | 17 - 18 | 2149 | 90 | ||
More Information
Set Membership: |
|

LOC4344163
Genomic Map