Assay Details
| Gene Symbol: LOC4344163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792552.2 | XP_015648038.1 | 18 - 19 | 2557 | 105 | |
| XM_015792553.1 | XP_015648039.1 | 18 - 19 | 2270 | 105 | ||
| GenBank mRNA | AK058967.1 | - | 6 - 7 | 861 | 105 | |
| AK064580.1 | - | 8 - 9 | 1183 | 105 | ||
| AK103870.1 | - | 18 - 19 | 2271 | 105 | ||
More Information
Set Membership: |
|

LOC4344163
Genomic Map