Assay Details
| Gene Symbol: LOC4344163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792552.2 | XP_015648038.1 | 1 - 2 | 642 | 157 | |
| XM_015792553.1 | XP_015648039.1 | 1 - 2 | 355 | 157 | ||
| GenBank mRNA | AK103870.1 | - | 1 - 2 | 356 | 157 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792552.2, XM_015792553.1 transcripts.
Set Membership: |
|

LOC4344163
Genomic Map