Assay Details
| Gene Symbol: LOC4344163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792552.2 | XP_015648038.1 | 21 - 22 | 2967 | 63 | |
| XM_015792553.1 | XP_015648039.1 | 21 - 22 | 2677 | 63 | ||
| GenBank mRNA | AK058967.1 | - | 9 - 10 | 1271 | 63 | |
| AK103870.1 | - | 21 - 22 | 2678 | 63 | ||
More Information
Set Membership: |
|

LOC4344163
Genomic Map