Assay Details
| Gene Symbol: LOC4344163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792552.2 | XP_015648038.1 | 7 - 8 | 1167 | 82 | |
| XM_015792553.1 | XP_015648039.1 | 7 - 8 | 880 | 82 | ||
| GenBank mRNA | AK103870.1 | - | 7 - 8 | 881 | 82 | |
More Information
Set Membership: |
|

LOC4344163
Genomic Map