Assay Details
| Gene Symbol: LOC4344210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792285.2 | XP_015647771.1 | 2 - 3 | 246 | 81 | |
| GenBank mRNA | AK243377.1 | - | 2 - 3 | 147 | 81 | |
| BT129027.1 | - | 2 - 3 | 164 | 81 | ||
| BT131351.1 | - | 1 - 2 | 69 | 81 | ||
| BT131357.1 | - | 2 - 3 | 135 | 81 | ||
| D01000.1 | - | 1 - 2 | 95 | 81 | ||
| EU325984.1 | - | 2 - 3 | 123 | 81 | ||
More Information
Set Membership: |
|

LOC4344210
Genomic Map