Assay Details
| Gene Symbol: LOC4344210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792285.2 | XP_015647771.1 | 4 - 5 | 439 | 110 | |
| GenBank mRNA | AK243377.1 | - | 4 - 5 | 340 | 110 | |
| BT129027.1 | - | 4 - 5 | 357 | 110 | ||
| BT131351.1 | - | 3 - 4 | 262 | 110 | ||
| BT131357.1 | - | 4 - 5 | 328 | 110 | ||
| D01000.1 | - | 3 - 4 | 288 | 110 | ||
| EU325984.1 | - | 4 - 5 | 316 | 110 | ||
More Information
Set Membership: |
|

LOC4344210
Genomic Map