Assay Details
| Gene Symbol: LOC4344210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792285.2 | XP_015647771.1 | 5 - 6 | 471 | 75 | |
| GenBank mRNA | AK243377.1 | - | 5 - 6 | 372 | 75 | |
| BT129027.1 | - | 5 - 6 | 389 | 75 | ||
| BT131351.1 | - | 4 - 5 | 294 | 75 | ||
| BT131357.1 | - | 5 - 6 | 360 | 75 | ||
| D01000.1 | - | 4 - 5 | 320 | 75 | ||
| EU325984.1 | - | 5 - 6 | 348 | 75 | ||
More Information
Set Membership: |
|

LOC4344210
Genomic Map