Assay Details
| Gene Symbol: LOC4344210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792285.2 | XP_015647771.1 | 7 - 8 | 604 | 85 | |
| GenBank mRNA | AK243377.1 | - | 7 - 8 | 505 | 85 | |
| BT129027.1 | - | 7 - 8 | 522 | 85 | ||
| BT131351.1 | - | 6 - 7 | 427 | 85 | ||
| BT131357.1 | - | 7 - 8 | 493 | 85 | ||
| D01000.1 | - | 6 - 7 | 453 | 85 | ||
| EU325984.1 | - | 7 - 8 | 481 | 85 | ||
More Information
Set Membership: |
|

LOC4344210
Genomic Map