Assay Details
| Gene Symbol: LOC4344440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794603.2 | XP_015650089.1 | 2 - 3 | 314 | 64 | |
| XM_015794604.2 | XP_015650090.1 | 2 - 3 | 465 | 64 | ||
| GenBank mRNA | AK120989.1 | - | 2 - 3 | 466 | 64 | |
More Information
Set Membership: |
|

LOC4344440
Genomic Map