Assay Details
| Gene Symbol: LOC4344440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794603.2 | XP_015650089.1 | 5 - 6 | 1506 | 138 | |
| XM_015794604.2 | XP_015650090.1 | 5 - 6 | 1657 | 138 | ||
| GenBank mRNA | AK120989.1 | - | 5 - 6 | 1658 | 138 | |
More Information
Set Membership: |
|

LOC4344440
Genomic Map