Assay Details
| Gene Symbol: LOC4344440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794603.2 | XP_015650089.1 | 7 - 8 | 2801 | 61 | |
| XM_015794604.2 | XP_015650090.1 | 7 - 8 | 2952 | 61 | ||
| GenBank mRNA | AK067468.1 | - | 2 - 3 | 1012 | 61 | |
| AK120989.1 | - | 7 - 8 | 2953 | 61 | ||
More Information
Set Membership: |
|

LOC4344440
Genomic Map