Assay Details
| Gene Symbol: LOC4344440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794603.2 | XP_015650089.1 | 9 - 10 | 3089 | 128 | |
| XM_015794604.2 | XP_015650090.1 | 9 - 10 | 3240 | 128 | ||
| GenBank mRNA | AK067468.1 | - | 4 - 5 | 1300 | 128 | |
| AK120989.1 | - | 9 - 10 | 3240 | 128 | ||
More Information
Set Membership: |
|

LOC4344440
Genomic Map