Assay Details
| Gene Symbol: LOC4344458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795474.2 | XP_015650960.1 | 3 - 4 | 1162 | 67 | |
| GenBank mRNA | AK064053.1 | - | 3 - 4 | 1165 | 67 | |
| AK101418.1 | - | 3 - 4 | 1164 | 67 | ||
More Information
Set Membership: |
|

LOC4344458
Genomic Map