Assay Details
| Gene Symbol: LOC4351327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764537.2 | XP_015620023.1 | 1 - 2 | 715 | 96 | |
| XM_015764539.2 | XP_015620025.1 | 1 - 2 | 708 | 96 | ||
| Gene Symbol: LOC4345283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793133.2 | XP_015648619.1 | 1 - 2 | 357 | 96 | |
| GenBank mRNA | AK120635.1 | - | 1 - 2 | 318 | 96 | |
More Information
Set Membership: |
|

LOC4351327
Genomic Map