Assay Details
| Gene Symbol: LOC4345313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795489.2 | XP_015650975.1 | 2 - 3 | 342 | 76 | |
| GenBank mRNA | AK068124.1 | - | 2 - 3 | 307 | 76 | |
More Information
Set Membership: |
|

LOC4345313
Genomic Map