Assay Details
| Gene Symbol: LOC4345339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795484.1 | XP_015650970.1 | 3 - 4 | 937 | 73 | |
| XM_015795485.2 | XP_015650971.1 | 3 - 4 | 833 | 73 | ||
| GenBank mRNA | AK071826.1 | - | 3 - 4 | 946 | 73 | |
| AK103906.1 | - | 3 - 4 | 817 | 73 | ||
| D83539.1 | - | 3 - 4 | 790 | 73 | ||
| EF122437.2 | - | 3 - 4 | 802 | 73 | ||
| FJ940194.1 | - | 3 - 4 | 853 | 73 | ||
More Information
Set Membership: |
|

LOC4345339
Genomic Map