Assay Details
| Gene Symbol: LOC4345339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795484.1 | XP_015650970.1 | 5 - 6 | 1221 | 118 | |
| XM_015795485.2 | XP_015650971.1 | 5 - 6 | 1117 | 118 | ||
| GenBank mRNA | AK071826.1 | - | 5 - 6 | 1230 | 118 | |
| AK103906.1 | - | 5 - 6 | 1101 | 118 | ||
| D83539.1 | - | 5 - 6 | 1074 | 118 | ||
| EF122437.2 | - | 5 - 6 | 1086 | 118 | ||
| FJ940194.1 | - | 5 - 6 | 1137 | 118 | ||
More Information
Set Membership: |
|

LOC4345339
Genomic Map