Assay Details
| Gene Symbol: LOC4345339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795484.1 | XP_015650970.1 | 9 - 10 | 1669 | 119 | |
| XM_015795485.2 | XP_015650971.1 | 9 - 10 | 1565 | 119 | ||
| GenBank mRNA | AK058551.1 | - | 1 - 2 | 125 | 119 | |
| AK071826.1 | - | 9 - 10 | 1678 | 119 | ||
| AK103906.1 | - | 9 - 10 | 1549 | 119 | ||
| D83539.1 | - | 9 - 10 | 1522 | 119 | ||
| EF122437.2 | - | 9 - 10 | 1534 | 119 | ||
| FJ940194.1 | - | 9 - 10 | 1585 | 119 | ||
More Information
Set Membership: |
|

LOC4345339
Genomic Map