Assay Details
| Gene Symbol: LOC4345414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792994.2 | XP_015648480.1 | 1 - 2 | 627 | 69 | |
| GenBank mRNA | AY224427.1 | - | 1 - 2 | 205 | 69 | |
More Information
Set Membership: |
|

LOC4345414
Genomic Map