Assay Details
| Gene Symbol: LOC4345424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793104.2 | XP_015648590.1 | 1 - 2 | 2616 | 166 | |
| GenBank mRNA | AK072348.1 | - | 1 - 2 | 2623 | 166 | |
More Information
Set Membership: |
|

LOC4345424
Genomic Map