Assay Details
| Gene Symbol: LOC4345446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795421.1 | XP_015650907.1 | 5 - 6 | 514 | 95 | |
| XM_015795422.1 | XP_015650908.1 | 4 - 5 | 484 | 95 | ||
| GenBank mRNA | AK061580.1 | - | 1 - 2 | 67 | 95 | |
More Information
Set Membership: |
|

LOC4345446
Genomic Map