Assay Details
| Gene Symbol: LOC4345446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795421.1 | XP_015650907.1 | 6 - 7 | 581 | 64 | |
| XM_015795422.1 | XP_015650908.1 | 5 - 6 | 551 | 64 | ||
| GenBank mRNA | AK061580.1 | - | 2 - 3 | 134 | 64 | |
| AK073566.1 | - | 1 - 2 | 334 | 64 | ||
More Information
Set Membership: |
|

LOC4345446
Genomic Map