Assay Details
| Gene Symbol: LOC4341799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788238.2 | XP_015643724.1 | 3 - 4 | 601 | 69 | |
| GenBank mRNA | AK070871.1 | - | 2 - 3 | 195 | 69 | |
More Information
Set Membership: |
|

LOC4341799
Genomic Map