Assay Details
| Gene Symbol: LOC4341799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788238.2 | XP_015643724.1 | 6 - 7 | 986 | 82 | |
| XM_015788239.2 | XP_015643725.1 | 6 - 7 | 847 | 82 | ||
| GenBank mRNA | AJ495798.1 | - | 2 - 3 | 149 | 82 | |
| AK070871.1 | - | 5 - 6 | 580 | 82 | ||
More Information
Set Membership: |
|

LOC4341799
Genomic Map