Assay Details
Gene Symbol: LOC4341799 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788238.2 | XP_015643724.1 | 7 - 8 | 1054 | 68 | |
XM_015788239.2 | XP_015643725.1 | 7 - 8 | 915 | 68 | ||
GenBank mRNA | AJ495798.1 | - | 3 - 4 | 217 | 68 | |
AK070871.1 | - | 6 - 7 | 648 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
