Assay Details
| Gene Symbol: LOC4341856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785598.2 | XP_015641084.1 | 4 - 5 | 1653 | 103 | |
| XM_015785599.1 | XP_015641085.1 | 3 - 4 | 793 | 103 | ||
| GenBank mRNA | AK121510.1 | - | 7 - 8 | 2045 | 103 | |
More Information
Set Membership: |
|

LOC4341856
Genomic Map