Assay Details
| Gene Symbol: LOC4342544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791587.2 | XP_015647073.1 | 10 - 11 | 1822 | 67 | |
| XM_015791588.1 | XP_015647074.1 | 7 - 8 | 1373 | 67 | ||
| GenBank mRNA | AK070264.1 | - | 10 - 11 | 1824 | 67 | |
More Information
Set Membership: |
|

LOC4342544
Genomic Map