Assay Details
| Gene Symbol: LOC4342544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791587.2 | XP_015647073.1 | 14 - 15 | 3228 | 109 | |
| XM_015791588.1 | XP_015647074.1 | 11 - 12 | 2779 | 109 | ||
| GenBank mRNA | AK070264.1 | - | 13 - 14 | 3316 | 109 | |
More Information
Set Membership: |
|

LOC4342544
Genomic Map