Assay Details
| Gene Symbol: LOC4342544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791587.2 | XP_015647073.1 | 5 - 6 | 703 | 90 | |
| XM_015791588.1 | XP_015647074.1 | 2 - 3 | 254 | 90 | ||
| GenBank mRNA | AK064551.1 | - | 2 - 3 | 206 | 90 | |
| AK070264.1 | - | 5 - 6 | 705 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015791588.1 transcripts.
Set Membership: |
|

LOC4342544
Genomic Map