Assay Details
| Gene Symbol: LOC4342620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791490.2 | XP_015646976.1 | 10 - 11 | 1336 | 56 | |
| XM_015791491.2 | XP_015646977.1 | 10 - 11 | 1290 | 56 | ||
| GenBank mRNA | AK071379.1 | - | 10 - 11 | 1289 | 56 | |
| AK103440.1 | - | 10 - 11 | 1283 | 56 | ||
More Information
Set Membership: |
|

LOC4342620
Genomic Map