Assay Details
| Gene Symbol: LOC4342620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791490.2 | XP_015646976.1 | 2 - 3 | 314 | 59 | |
| XM_015791491.2 | XP_015646977.1 | 2 - 3 | 268 | 59 | ||
| GenBank mRNA | AK071379.1 | - | 2 - 3 | 267 | 59 | |
| AK103440.1 | - | 2 - 3 | 261 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015791490.2, XM_015791491.2 transcripts.
Set Membership: |
|

LOC4342620
Genomic Map