Assay Details
| Gene Symbol: LOC4342620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791490.2 | XP_015646976.1 | 6 - 7 | 942 | 94 | |
| XM_015791491.2 | XP_015646977.1 | 6 - 7 | 896 | 94 | ||
| GenBank mRNA | AK071379.1 | - | 6 - 7 | 895 | 94 | |
| AK103440.1 | - | 6 - 7 | 889 | 94 | ||
More Information
Set Membership: |
|

LOC4342620
Genomic Map