Assay Details
| Gene Symbol: LOC4342787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792405.2 | XP_015647891.1 | 3 - 4 | 686 | 73 | |
| XM_015792406.2 | XP_015647892.1 | 3 - 4 | 690 | 73 | ||
| GenBank mRNA | DQ374663.1 | - | 3 - 4 | 487 | 73 | |
More Information
Set Membership: |
|

LOC4342787
Genomic Map