Assay Details
| Gene Symbol: LOC4342787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792405.2 | XP_015647891.1 | 6 - 7 | 968 | 152 | |
| XM_015792406.2 | XP_015647892.1 | 6 - 7 | 972 | 152 | ||
| GenBank mRNA | AK111874.1 | - | 6 - 7 | 968 | 152 | |
| DQ374663.1 | - | 6 - 7 | 769 | 152 | ||
More Information
Set Membership: |
|

LOC4342787
Genomic Map