Assay Details
| Gene Symbol: LOC4342787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792405.2 | XP_015647891.1 | 9 - 10 | 1308 | 62 | |
| GenBank mRNA | AK111874.1 | - | 9 - 10 | 1308 | 62 | |
| DQ374663.1 | - | 9 - 10 | 1109 | 62 | ||
More Information
Set Membership: |
|

LOC4342787
Genomic Map