Assay Details
| Gene Symbol: LOC4342894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789515.2 | XP_015645001.1 | 6 - 7 | 617 | 89 | |
| XM_015789516.2 | XP_015645002.1 | 6 - 7 | 618 | 89 | ||
| XM_015789517.1 | XP_015645003.1 | 6 - 7 | 615 | 89 | ||
| XM_015789518.1 | XP_015645004.1 | 6 - 7 | 612 | 89 | ||
| GenBank mRNA | AK058966.1 | - | 6 - 7 | 620 | 89 | |
| AK068904.1 | - | 6 - 7 | 618 | 89 | ||
More Information
Set Membership: |
|

LOC4342894
Genomic Map