Assay Details
| Gene Symbol: LOC4343922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789510.2 | XP_015644996.1 | 2 - 3 | 298 | 93 | |
| GenBank mRNA | AK074019.1 | - | 2 - 3 | 247 | 93 | |
More Information
Set Membership: |
|

LOC4343922
Genomic Map