Assay Details
| Gene Symbol: LOC4343954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791568.2 | XP_015647054.1 | 1 - 2 | 411 | 82 | |
| XM_015791569.2 | XP_015647055.1 | 1 - 2 | 408 | 82 | ||
| GenBank mRNA | AK058506.1 | - | 1 - 2 | 367 | 82 | |
| AK069142.1 | - | 1 - 2 | 395 | 82 | ||
| AK103285.1 | - | 1 - 2 | 289 | 82 | ||
More Information
Set Membership: |
|

LOC4343954
Genomic Map