Assay Details
| Gene Symbol: LOC4343971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789227.2 | XP_015644713.1 | 6 - 7 | 950 | 99 | |
| GenBank mRNA | AK288055.1 | - | 6 - 7 | 924 | 99 | |
More Information
Set Membership: |
|

LOC4343971
Genomic Map