Assay Details
| Gene Symbol: LOC4344699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794957.2 | XP_015650443.1 | 10 - 11 | 1945 | 87 | |
| GenBank mRNA | AK106701.1 | - | 10 - 11 | 1129 | 87 | |
More Information
Set Membership: |
|

LOC4344699
Genomic Map