Assay Details
Gene Symbol: LOC4344699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794957.2 | XP_015650443.1 | 4 - 5 | 1407 | 59 | |
GenBank mRNA | AK106701.1 | - | 4 - 5 | 591 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
