Assay Details
Gene Symbol: LOC4344699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794957.2 | XP_015650443.1 | 8 - 9 | 1798 | 113 | |
GenBank mRNA | AK106701.1 | - | 8 - 9 | 982 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
