Assay Details
| Gene Symbol: LOC4344803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794252.2 | XP_015649738.1 | 3 - 4 | 210 | 89 | |
| GenBank mRNA | AK121665.1 | - | 3 - 4 | 197 | 89 | |
More Information
Set Membership: |
|

LOC4344803
Genomic Map